STERIPLAN™ Petri Dish, from Soda-lime Glass
STERIPLAN™ Petri Dish, from Soda-lime Glass
Actual product may vary
Fisherbrand
STERIPLAN™ Petri Dish, from Soda-lime Glass
Petri dishes are used for the cultivation of bacteria, yeasts, plant and animal cells on solid media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Height (Metric)
FKS1174083460 mm15 mm
FKS11750844100 mm15 mm
Catalog number FKS11740834
Price (SGD)
80.00
Pack
Add to cart
Diameter (Metric):
60 mm
Height (Metric):
15 mm
Price (SGD)
80.00
Pack
Add to cart
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow.
  • Free from bubbles and streaks
  • Even distribution of sewing bottom possible
Specifications
AutoclavableNot Autoclavable
MaterialSoda Lime Glass
For Use With (Application)Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; Cell culture
No. per Pack10/Pk.
Height (Metric)15 mm
Diameter (Metric)60 mm
Unit SizePack