Check Order Status
Silver Filter
Actual product may vary
Fisherbrand
Silver Filter
Have Questions?
Catalog Number
Diameter (Metric)
FKS11740019
47 mm
Catalog number
FKS11740019
Price (SGD)
-
Diameter (Metric):
47 mm
Specifications
Airflow Rate
350L/cm
2
Bubble Point
13psi
Material
Silver
Pore Size
0.2 μm
Quantity
25Pack
Type
Filter
Diameter (Metric)
47 mm