Grade 926 Sterilization Paper
Grade 926 Sterilization Paper
Actual product may vary
Fisherbrand
Grade 926 Sterilization Paper
Used as wrap for medical clothes.
Have Questions?
Catalog NumberDimensions (L x W)
FKS1173851350 x 50 cm
Catalog number FKS11738513
Price (SGD)
346.00
PK
Add to cart
Dimensions (L x W):
50 x 50 cm
Price (SGD)
346.00
PK
Add to cart
  • Weight: 60g/m2
  • Creped paper
Applications:
  • Used in steam, gas, gamma-ray sterilization
Specifications
AutoclavableAutoclavable
Certifications/ComplianceEN 868-2
ColorGreen
For Use With (Application)Wrapping of medical clothes, surgical instruments and others articles used in steam, gas and gamma-ray sterilization
MaterialPaper
Quantity500 Pk.
Dimensions (L x W)50 x 50 cm
Length (Metric)50 cm
Width (Metric)50 cm
Weight (Metric)60 gsm
Product TypeSterilization Paper
Grade926
Unit SizePK