Check Order Status
UVL-56 Handheld UV Lamp
Fisherbrand
UVL-56 Handheld UV Lamp
Use these lamps for variety of applications including fluorescence of stained gels and sterilization. UVP™ Handheld UV Lamps are made of durable, lightweight plastic with an ergonomically designed handle.
Have Questions?
Catalog number
FKS11738221
Price (SGD)
1,419.00
EA
Add to cart
Price (SGD)
1,419.00
EA
Add to cart
Comfortable, ergonomically designed handle
On/off switch is located on the back of the lamp
Lamps are lightweight for easy handling
Durable plastic housing
Six watts
Specifications
Light Source
UV-A
Voltage
230 V
Wavelength
365 nm
Case Material
Plastic
Dimensions (L x W x H)
378 x 81 x 64 mm
Model
UVL-56 Handheld UV Lamp