Orion™ ISE Calibration Standards
Orion™ ISE Calibration Standards
Actual product may vary
Fisherbrand
Orion™ ISE Calibration Standards
Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
FKS117376771000 ppm as CaCO3
FIS13-641-924C1000 ppm as N
FIS13-641-8110.1 M
FIS13-641-7680.1 M NaHCO3
FKS117276770.1 M NaHCO3
FIS13-641-8760.1 M NaCl
FIS13-299-871000 ppm as Cl-
FIS13-641-872100 ppm as F-
FKS102529212 ppm as F- with TISAB II
FIS13-641-9170.1 M KCl
FIS13-641-7471000 ppm as Na+
FIS13-641-862100 ppm as CaCO3
Catalog number FKS11737677
Price (SGD)
-
Concentration:
1000 ppm as CaCO3
Take accurate, reproducible ion concentration measurements by calibrating your Ion Selective Electrode with high-quality Thermo Scientific™ Orion™ ISE Calibration Standards. Use these standards with electrodes including ammonia, chloride, fluoride, nitrate, potassium, silver, sodium and sulfide. Orion calibration standards are NIST traceable and manufactured under ISO 9001 quality standards.
Use the Orion ISE calibration standards with our extensive line of ion selective electrodes (ISEs) for ammonia, ammonium, bromide, cadmium, calcium, carbon dioxide, chloride, chlorine, copper, cyanide, fluoride, iodide, lead, nitrate, potassium, silver, sodium and sulfide analysis. Whether you are in the lab or in the field, you can be confident that Orion solutions will deliver accurate and repeatable measurements. Orion solutions are produced in controlled batches using ultrapure water and undergo rigorous quality testing throughout the process to prevent contamination. Our strict quality procedures verify correct bottle labeling and volume, so each batch meets specific lot release criteria. Bottles are sealed and torque-tested to ensure bottle caps are secure and solutions remain free of contamination. Rigorous lifetime testing ensures sealed Orion solutions remain within published specifications for their entire shelf life and do not degrade over time.
* Orion calibration standards are NIST traceable.
* Orion calibration standards are manufactured under ISO 9001 quality standards.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionCarbon Dioxide Standard
Packaging1 Bottle
Certificate of AnalysisYes
Concentration or Composition NotesC of A Acceptance Range: 0.01000 M +/- 0.00005 M Sodium Bicarbonate
Traceability to NISTYes
Product LineCarbon Dioxide Ion Selective Electrodes Including 9502BNWP
Physical FormLiquid
Concentration1000 ppm as CaCO3
ColorColorless
For Use With (Equipment)Carbon Dioxide Ion Selective Electrode
Solution TypeCalibration Standard
TypeCalibration Standard
Quantity475 mL
Unit SizeEA