AR-Glass Graduated Pipet
AR-Glass Graduated Pipet
Actual product may vary
Fisherbrand
AR-Glass Graduated Pipet
Provides volumetric accuracy for your pipetting needs. Hirschmann™ Clear Glass Graduated Pipettes with Amber Stain Graduation with a cotton plug end as standard, short form for tissue culture.
Have Questions?
Catalog NumberVolume (Metric)Graduations
FKS1173649210 mL0.1 mL
Catalog number FKS11736492
Price (SGD)
408.00
PK
Add to cart
Volume (Metric):
10 mL
Graduations:
0.1 mL
Price (SGD)
408.00
PK
Add to cart
  • Main point ring graduation
  • Cotton plug end
  • Class AS, Conformity certified, with dated batch
Specifications
DescriptionShort form for tissue culture
Diameter (Metric) Outer8 mm
MaterialSoda Lime Glass
For Use With (Application)For tissue culture and working in cleanroom benches
GraduatedYes
Graduation ColorAmber
Graduations0.1 mL
Graduation TypeDurable amber stain diffusion ink
Quantity12 Pack
Volume (Metric)10 mL
Length (Metric)230 ±3 mm
TypeGraduated Pipet
Unit SizePK