Microspatula Set
Microspatula Set
Actual product may vary
Fisher Catalogue
Microspatula Set
Set of four with plastic, color-coded handles
Have Questions?
Catalog NumberLength (Metric)
FKS117343566.35 cm
Catalog number FKS11734356
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
6.35 cm
Set of four with plastic, color-coded handles
  • Dispense and transfer samples during weighing
  • Apply micro amounts of adhesive for tacking down specimens, spreading paste and mixing granular samples
  • Flat-ended stainless-steel blades
  • Four sizes of blades: 0.1, 0.3, 0.5, and 1.0mm
  • Overall length: 2.5 in. (6cm)
Specifications
Product TypeMicrospatula Set
For Use With (Equipment)Balances, microscopes and weighing boats
For Use With (Application)Designed to dispense and transfer samples during weighing, can also be used for tacking down specimens, spreading paste and mixing granular samples by apply micro amounts of adhesive
Length (English)2.5 in.
Length (Metric)6.35 cm
Unit SizeST