Indoor/Outdoor Permanent Adhesive Vinyl Label Cartridges
Indoor/Outdoor Permanent Adhesive Vinyl Label Cartridges
Actual product may vary
Indoor/Outdoor Permanent Adhesive Vinyl Label Cartridges
Indoor/Outdoor Permanent Adhesive Vinyl Label Cartridges
Fisherbrand
Indoor/Outdoor Permanent Adhesive Vinyl Label Cartridges
Indoor/Outdoor Permanent Adhesive Vinyl Label Cartridges adhere to irregular, rough and curved surfaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (L x W)
FKS11733484Black on Orange21 x 0.75 in. (6.4 m x 19.05 mm)
FIS19-102-781Black on White21 x 0.75 in. (6.4 m x 19.05 mm)
Catalog number FKS11733484
Price (SGD)
177.00
BOX
Add to cart
Color:
Black on Orange
Dimensions (L x W):
21 x 0.75 in. (6.4 m x 19.05 mm)
Price (SGD)
177.00
BOX
Add to cart
Adheres to irregular, rough and curved surfaces
  • For use with BMP™21, BMP21-LAB, BMP21-PLUS, IDPal™, LabPal™ Printers (M210, M210-LAB, M211, BMP21-PLUS, BMP21-LAB, IDPAL, LABPAL).
  • Made of 8-10 year indoor/outdoor material.
  • Matte finish with permanent adhesive.
  • Stands up to grease, oil and most industrial chemicals.
  • Available in various sizes and colors.
Applications:
General identification, laboratory identification, panel identification, panel labels, plant labels, safety labels, Lean/5S and visual workplace labels, pipe marking, tool labeling, equipment labeling and outdoor labeling.
Specifications
Product TypeLabel Cartridge
ColorBlack on Orange
For Use With (Equipment)Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer, BMP21-LAB Label Printer, BMP21-PLUS Label Printer
MaterialVinyl
Print TechnologyThermal Transfer
Length (English)21 ft.
Width (English)0.75 in.
Length (Metric)6.4 m
Width (Metric)19.5 mm
Thickness (English)0.004 in.
Thickness (Metric)0.102 mm
Dimensions (L x W)21 x 0.75 in. (6.4 m x 19.05 mm)
Adhesive TypePermanent Acrylic
DescriptionVinyl tape for BMP21-PLUS; BMP21-LAB; BMP21; IDPAL; LABPAL
FinishGloss
FormatContinuous
Temperature Range (English)-40°F to +179.6°F
Temperature Range (Metric)-40°C to +82°C
Certifications/ComplianceBS5609:1986 Section 2 and section 3
For Use With (Application)Freezer
Quantity1 Ea.
Alternate Part NumberM21-750-595-OR
Unit SizeBOX