Orion™ ROSS Ultra™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode
Orion™ ROSS Ultra™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode
Actual product may vary
Fisherbrand
Orion™ ROSS Ultra™ Flat Surface Refillable Epoxy Bodied Combination pH Electrode
Measure flat, soft, moist, semi-solid or small volume samples with epoxy body flat surface pH electrodes.
Have Questions?
Catalog NumberConnector Types
FKS11732767BNC (Waterproof)
Catalog number FKS11732767
Price (SGD)
2,131.00
EA
Add to cart
Connector Types:
BNC (Waterproof)
Price (SGD)
2,131.00
EA
Add to cart
Thermo Scientific™ Orion™ ROSS Ultra Flat Surface Electrodes have a uniquely designed, flat sensing bulb that allow for reliable pH measurement on samples such as paper, textiles, skin, meat, cheese, or in agar gel plates.
Orion™ ROSS Ultra Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonomous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
Solutions required 
Electrode fill solution
810007     ROSS pH electrode fill solution, 5 x 60 mL bottles
Electrode storage solution
810001     ROSS pH electrode storage solution, 475 mL bottle
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
810199     All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
                 electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
                 storage bottle
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with 
810007     ROSS pH electrode fill solution 60 mL 1 bottle
Warranty period
24 months from date of purchase
Specifications
DescriptionUltra pH Electrode
Connector TypesBNC (Waterproof)
JunctionGlass Fiber
MaterialEpoxy Body
pH Range0 to 14
Temperature (Metric) Operating0°C to 100°C
TypepH Electrode
ElectrodeFlat surface tip epoxy body combination electrode; 20 x 12mm
Filling Solution Type3M KCl
Internal ReferenceROSS
Precision0.01
Temperature (English) Operating32°F to 212°F
Warranty2 Years
Length (English) Cable39 in.
Length (Metric)1 m
Unit SizeEA