Colored ABS 50 Place Slide Boxes
Colored ABS 50 Place Slide Boxes
Actual product may vary
Colored ABS 50 Place Slide Boxes
Colored ABS 50 Place Slide Boxes
Fisherbrand
Colored ABS 50 Place Slide Boxes
Store and transport 50 slides safely with durable ABS cases. Fisherbrand™ Colored ABS 50 Place Slide Boxes keep standard microscope slides in numbered, grooved slots. It also provides safe, dust-free storage.
Have Questions?
Catalog NumberColor
FKS11731486Blue
Catalog number FKS11731486
Price (SGD)
35.00
EA
Add to cart
Color:
Blue
Price (SGD)
35.00
EA
Add to cart
  • Molded, rigid box securely holds slides in grooved slots
  • Durable ABS case with cork lining
  • Lining and slots are numbered to correspond to slide inventory sheet
  • Easily open case with hinged cover featuring a thumb latch
  • Stackable cases include feet for maximum stability
Specifications
TypeSlide Storage Box
Slide Capacity50
Closure TypeThumb Latch
ColorBlue
Device Holds50 Microscope Slides (25 x 75 mm or 1 x 3 in.)
Dimensions (L x W x H)209 x 86 x 35 mm
For Use With (Equipment)Standard (25 x 75 mm or 1 x 3 in.) Microscope Slides
Height (English)1.4 in.
Height (Metric)35 mm
IncludesSlide Inventory Sheet
Length (English)8.2 in.
Length (Metric)209 mm
MaterialABS Plastic with Cork Lining
ShapeRectangular
StackableYes
Width (English)3.4 in.
Width (Metric)86 mm
Unit SizeEA