Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
FKS11730884 | 250 mL |
FKS11351465 | 1000 mL |
Beakers are cylindrical vessels with a straight side wall. Thanks to this shape, beakers are universally usable laboratory vessels and usually serve as cooking glasses, collecting vessels and for preparing, mixing and heating solutions. Easy filling is possible via the large vessel opening. Beakers are available in various shapes and sizes: high and low, with or without spout, with or without scale.