Check Order Status
Quartz Microplates
Actual product may vary
Fisherbrand
Quartz Microplates
Provides for chemical resistance, UV transmission, microscopic observation and thermal stability. Hellma™ Quartz Microplates offer many advantages over their plastic counterparts.
Have Questions?
Catalog Number
Color
FKS11729655
Clear
Catalog number
FKS11729655
Price (SGD)
18,999.00
EA
Add to cart
Color:
Clear
Price (SGD)
18,999.00
EA
Add to cart
96 wells
Even well base, less than the wavelength of light
Each well completely liquid guaranteed
Specifications
Format
Standard
Array
8 x 12
Color
Clear
Product Type
96-well Microplate
Material
Synthetic Quartz Glass
No. of Wells
96
Total Volume (Metric) Well
300 μL
Quantity
Each
Depth (Metric)
12.58 mm
Diameter (Metric) Well
6.6 mm