Check Order Status
Grade 122 General Purpose Filter Papers
Actual product may vary
Fisherbrand
Grade 122 General Purpose Filter Papers
A general purpose filter paper used for all general works in the laboratory for excellent and consistent filtration.
Have Questions?
Catalog Number
Format
Diameter (Metric)
FKS11728752
Disc
150 mm
Catalog number
FKS11728752
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Format:
Disc
Diameter (Metric):
150 mm
Very fast filtration velocity
Available in a range of sizes
Available as plain or folded discs
Applications include hospitals, research laboratories, schools, chemical industries, pharmaceuticals and more
Specifications
Format
Disc
Material
Cellulose
Particle Retention
17 to 30 μm
Pore Size
17 to 30 μm
For Use With (Application)
For all general works in laboratories
Grade
122
Quantity
100/Pk.
Thickness (Metric)
0.17 mm
Diameter (Metric)
150 mm