Tamper Evident Screw Cap, Wide Mouth, Blue PP, for Soda-Lime Glass Screw-Top Square Bottles
Tamper Evident Screw Cap, Wide Mouth, Blue PP, for Soda-Lime Glass Screw-Top Square Bottles
Actual product may vary
Tamper Evident Screw Cap, Wide Mouth, Blue PP, for Soda-Lime Glass Screw-Top Square Bottles
Tamper Evident Screw Cap, Wide Mouth, Blue PP, for Soda-Lime Glass Screw-Top Square Bottles
Fisherbrand
Tamper Evident Screw Cap, Wide Mouth, Blue PP, for Soda-Lime Glass Screw-Top Square Bottles
Tamper evident cap for wide mouth/short form DIN soda lime glass bottle thread.
Have Questions?
Catalog NumberThread Size (Metric)
FKS11728011GL 32
Catalog number FKS11728011
Price (SGD)
43.00
PK
Add to cart
Thread Size (Metric):
GL 32
Price (SGD)
43.00
PK
Add to cart
Tamper evident, blue polypropylene, linerless screw cap for wide mouth/short form DIN thread soda lime glass bottles.
  • Tamper evident ring
  • LInerless
  • Short form DIN thread
Specifications
For Use With (Application)Storage of samples and reagents; Sample collection; Transport and shipping
MaterialPolypropylene
Quantity10/Pk.
Thread Size (Metric)GL 32
AutoclavableAutoclavable
Diameter (Metric)44 mm
Unit SizePK