Stainless Steel Graduated Pouring Beakers
Stainless Steel Graduated Pouring Beakers
Actual product may vary
Fisherbrand
Stainless Steel Graduated Pouring Beakers
Easily measure and pour samples with Stainless Steel Graduated Pouring Beakers.
Have Questions?
Catalog NumberCapacity (Metric)
FKS11727964500 mL
Catalog number FKS11727964
Price (SGD)
684.00
EA
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
684.00
EA
Add to cart
  • Measure volumes easily with internal graduation markers
  • Great for higher temperatures— withstand temperatures up to 650°F (182°C)
  • Features easy grip handles for convenient pouring
  • Beakers are graduated in mL and oz.
  • Constructed of medium gauge 304 stainless steel
Specifications
Capacity (English)16.9 oz.
Diameter (English) Outer3.5 in.
Diameter (Metric) Outer90 mm
Capacity (Metric)500 mL
MaterialStainless Steel
Height (English)4.5 in.
Height (Metric)115 mm
Temperature (English) Max. Operating800.1°F
Temperature (Metric) Max. Operating426.7°C
TypeBeaker
SterilitySterile
GraduatedYes
StyleHandled
Unit SizeEA