C-Shape Vinyl Coated Lead Rings
C-Shape Vinyl Coated Lead Rings
Actual product may vary
Fisherbrand
C-Shape Vinyl Coated Lead Rings
Stabilize vessels and labware or immerse in water baths. Fisherbrand™ Vinyl-coated Lead Rings keep flasks and other labware weighted down without causing damage.
Have Questions?
Catalog NumberDiameter (Metric) InnerDiameter (Metric) OuterWeight (Metric)Size
FKS1172672542 mm75 mm0.277 kg125 to 500 mL
Catalog number FKS11726725
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric) Inner:
42 mm
Diameter (Metric) Outer:
75 mm
Weight (Metric):
0.277 kg
Size:
125 to 500 mL
  • Vinyl-coated lead rings will not mark or scratch benchtops and labware
  • C-shape format allows some manual size variation for a customized fit
Specifications
Product TypeC-shape
MaterialLead
Quantity1 Each
Size125 to 500 mL
Diameter (Metric) Outer75 mm
Diameter (Metric) Inner42 mm
Weight (Metric)0.277 kg
Unit SizeEA