DURAN™ Watch Glass Dish, fused rim
DURAN™ Watch Glass Dish, fused rim
Actual product may vary
DURAN™ Watch Glass Dish, fused rim
DURAN™ Watch Glass Dish, fused rim
DURAN™ Watch Glass Dish, fused rim
DURAN™ Watch Glass Dish, fused rim
Fisherbrand
DURAN™ Watch Glass Dish, fused rim
The concave shape of the watch glass dish allow working in very small quantities (e.g. for pretreatments) and covering laboratory glassware with wide openings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS11726602100 mm
FKS1170660260 mm
Catalog number FKS11726602
Price (SGD)
610.00
PK
Add to cart
Diameter (Metric):
100 mm
Price (SGD)
610.00
PK
Add to cart
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Watch glasses, for example, are concave curved. The small trough created in this way allows liquids or solids to be filled in.
  • Reduced risk of injury due to fused rim
Specifications
AutoclavableAutoclavable
MaterialBorosilicate Glass (3.3)
For Use With (Application)Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass
No. per Pack10/Pk.
Diameter (Metric)100 mm
Unit SizePK