Borosilicate Crystallizing Dishes with Spout
Borosilicate Crystallizing Dishes with Spout
Actual product may vary
Fisherbrand
Borosilicate Crystallizing Dishes with Spout
Includes spout. DURAN™ BoroSilicate Crystallizing Dishes with Spout have good thermal shock resistance.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FKS11726582900 mL140 mm75 mm
Catalog number FKS11726582
Price (SGD)
1,090.00
PK
Add to cart
Capacity (Metric):
900 mL
Diameter (Metric):
140 mm
Height (Metric):
75 mm
Price (SGD)
1,090.00
PK
Add to cart
  • Good thermal-shock resistance
  • High operating temperature (+500 °C)
  • Not designed for use under differential pressure or vacuum conditions.
Specifications
Bottom ShapeFlat
Capacity (Metric)900 mL
MaterialBorosilicate Glass
ShapeRound
TypeEvaporating Dish
Height (Metric)75 mm
Diameter (Metric)140 mm
Unit SizePK