1x1 image pixel for data collection
ChemFluor™ PTFE Tubing
ChemFluor™ PTFE Tubing
Actual product may vary
Fisherbrand
ChemFluor™ PTFE Tubing
Ideal for chemical transfer and pharmaceutical processes
Have Questions?
Catalog NumberDiameter (Metric) Inner
FKS117258984 mm
Catalog number FKS11725898
Price (SGD)
3,057.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Diameter (Metric) Inner:
4 mm
Price (SGD)
3,057.00
EA
Add to cart
  • Most flexible fluoropolymer.
  • Excellent chemical compatibility.
  • Translucent.
  • Certification: FDA compliant.
  • Temperature range: -450 to +550°F (-268 to +288°C).
  • Sterilize: Autoclave, ethylene oxide, or dry heat.
Specifications
Certifications/ComplianceEuropean Pharmacopoeia;FDA;3A;NSF-51;ADCF;NSF-61;USP;Non-DEHP;Non-Phthalate
DescriptionMost flexible fluoropolymer.
AutoclavableAutoclavable
Diameter (Metric) Inner4 mm
MaterialPTFE
Diameter (Metric) Outer6.4 mm
Overall Length (English)50 ft.
Overall Length (Metric)15.2 m
Thickness (Metric)1.19 mm
For Use With (Application)Chemical transfer and pharmaceutical processes.
Min. Operating Temperature (English)-450°F
Min. Operating Temperature (Metric)-267°C
Packaging Quantity50 ft./Pk.
Resistance PropertiesExcellent chemical compatibility.
Sterilization MethodAutoclave, ethylene oxide, or dry heat.
ColorTranslucent
Maximum Pressure117 psi
Max. Operating Temperature (English)550°F
Max. Operating Temperature (Metric)287°C
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link