DURAN™ Original Laboratory GL Bottle Screw Cap, Linerless Lip Seal, PP
DURAN™ Original Laboratory GL Bottle Screw Cap, Linerless Lip Seal, PP
Actual product may vary
Fisherbrand
DURAN™ Original Laboratory GL Bottle Screw Cap, Linerless Lip Seal, PP
Screw cap with reliable plug seal.
Have Questions?
Catalog NumberThread Size (Metric)Color
FKS11725434GL 45Yellow
Catalog number FKS11725434
Price (SGD)
84.00
PK
Add to cart
Thread Size (Metric):
GL 45
Color:
Yellow
Price (SGD)
84.00
PK
Add to cart
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
  • Cap with integral lip seal provides effective sealing for short term storage
  • Pouring ring for drip-free pouring
  • The colour helps prevent interchanging of cap mix-ups and reduces the risk of cross contamination
  • Fully compatible with all the GL 45 threads and therefore highly versatile
Specifications
ColorYellow
DescriptionScrew cap
For Use With (Application)Autoclaving media and reagents; Short term sample and media storage; color coding and identification
MaterialPolypropylene
Quantity10/Pk.
Thread Size (Metric)GL 45
AutoclavableAutoclavable
Diameter (Metric)54 mm
Unit SizePK