Cellulose Membranes
Cellulose Membranes
Actual product may vary
Fisherbrand
Cellulose Membranes
Suitable for Clarification and sterilizing filtration of biological fluids . Fisherbrand™ Cellulose Membranes filters are composed of pure cellulose acetate modified to offer researchers the lowest binding filters available.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Bubble Point
FKS1172531547 mm3.1bar
FKS1170531547 mm4bar
Catalog number FKS11725315
Price (SGD)
331.00
PK
Add to cart
Diameter (Metric):
47 mm
Bubble Point:
3.1bar
Price (SGD)
331.00
PK
Add to cart
  • Filtration sterile of aqueous solutions (nutrient media, sera, buffers)
  • Sterilizing filtration of proteins and enzymes
  • Provide high throughputs
  • Reduces filter changes when filtering proteinaceous solutions
  • Has Receptor binding studies
  • Low binding characteristics
  • Made of hydrophilic membranes
  • Uniform pore size structure allows consistent flow rates
  • Present strength and dimension stability (130psi) and resist to 135°C
Specifications
Bubble Point3.1bar
ColorWhite
Filter TypeMicrofilter
MaterialCellulose Acetate
Pore Size0.45 μm
SterilityNon-sterile
Flow Rate40mL/min./cm2
For Use With (Application)Sterilizing filtration of aqueous solutions (Nutrient media, Buffers and sera), sterilizing filtration of Proteins and enzyme, receptor binding studies
Gas Flow Rate25L/min./cm2
HydrophobicityHydrophilic
Quantity100 Pack
TypePlain Disc
WettabilityYes
Diameter (Metric)47 mm
Unit SizePK