Traceable™ Dew-Point/Wet-Bulb/Humidity/Thermometer
Traceable™ Dew-Point/Wet-Bulb/Humidity/Thermometer
Actual product may vary
Fisherbrand
Traceable™ Dew-Point/Wet-Bulb/Humidity/Thermometer
Traceable™ Dew-Point/Wet-Bulb/Humidity/Thermometer shows current, minimum and maximum displays.
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS11724196-40°C to +70°C
Catalog number FKS11724196
Price (SGD)
319.00
EA
Add to cart
Temperature Range (Metric):
-40°C to +70°C
Price (SGD)
319.00
EA
Add to cart
  • Set high/low alarm for all four readings (dew-point, wet-bulb, humidity and temperature)
  • Alarm may be set in 0.1° increments; sounds for one minute, and red LED flashes until readings return to nonalarm conditions
  • Temperature range: -40° to +70°C and -40° to +158°F
  • Temperature resolution: 0.1° (below 100°)
  • Accuracy: ±1°C
  • Relative humidity range: 1 to 99%
  • Resolution: 0.1% RH
  • Accuracy: ±4% between 20 to 80% RH
  • Dimensions: 4.13 x 4.13 x 1 in. (10.5 x 10.5 x 2.5 cm)
  • LCD digits: 1 in. H (2.5 cm)
  • Weight: 4.25 oz. (0.12 kg)
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Dimensions (L x W x H)4.12 x 4.13 x 1 in. (10.5 x 10.5 x 2.5 cm)
Display Dimensions1 in. H
For Use With (Application)Monitoring current, minimum and maximum displays
Height (English)1 in.
Height (Metric)2.5 cm
IncludesTwo AA alkaline batteries for one year of continuous monitoring, High-impact, chemical-resistant ABS plastic case
Length (Metric)105 mm
MaterialABS Plastic
Resolution0.1°C
Weight (English)4.25 oz.
Weight (Metric)120 g
Width (English)4.13 in.
Product TypeTraceable Thermometer
Length (English)4.12 in.
Max. Temperature (English)158°F
Max. Temperature (Metric)70°C
Min. Temperature (English)-40°F
Min. Temperature (Metric)-40°C
Temperature Range (English)-40°F to +158°F
Temperature Range (Metric)-40°C to +70°C
CalibratedYes
Accuracy±1°C
DisplayDigital LCD
Width (Metric)10.5 cm
Unit SizeEA