Grade 51 Ashless Hardened Quantitative Filter Paper
Grade 51 Ashless Hardened Quantitative Filter Paper
Actual product may vary
Fisherbrand
Grade 51 Ashless Hardened Quantitative Filter Paper
Used for quantitative analysis.
Have Questions?
Catalog NumberDiameter (Metric)
FKS1172237570 mm
Catalog number FKS11722375
Price (SGD)
-
Diameter (Metric):
70 mm
  • Plain disc, wet strengthened ashless filter paper
  • Made with 100% cotton linters fibers
  • Paper have been washed out with acid
  • Finally cleaned with demineralized water
  • Ash content: 0.007%
Applications:
  • Used for analysis of acid solution and pressure filtration
  • Food industry
  • Beverage industry
  • Atmospheric analysis
Specifications
DescriptionAsh Content: <0.008%
FormatDisc
MaterialCotton Linters
Particle Retention12 to 15 μm
Pore Size12 to 15 μm
Filtration Speed10 sec.
For Use With (Application)Acid solutions analysis and for filtration under high pressure
Grade51
Quantity100 Pk.
Thickness (Metric)0.18 mm
Diameter (Metric)70 mm
Unit SizePK