1x1 image pixel for data collection
Premium Microscope Slide Box, 100 Place
Premium Microscope Slide Box, 100 Place
Actual product may vary
Fisherbrand
Premium Microscope Slide Box, 100 Place
Store and transport 100 slides safely with durable ABS cases. Colored ABS 100 Place Slide Boxes keep standard microscope slides in numbered, grooved slots. Provides safe, dust-free storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorLining Material
FKS11721486WhiteFoam
FIS03-448-1BlueFoam
FKS11701486BlueFoam
FIS03-448-2GreenFoam
FIS03-448-4YellowFoam
FIS03-448-5WhiteFoam
FIS03-448-3RedFoam
FKS11711486RedFoam
Catalog number FKS11721486
Price (SGD)
24.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Color:
White
Lining Material:
Foam
Price (SGD)
24.00
EA
Add to cart
Store and transport 100 slides safely with durable ABS cases. Fisherbrand™ Colored ABS 100 Place Slide Boxes keep standard microscope slides in numbered, grooved slots. Provides safe, dust-free storage.
  • Molded, rigid boxes securely hold standard microscope slides in grooved slots.
  • Durable ABS case available lined lined with foam or unlined.
  • Slots and lining are numbered to correspond to slide inventory sheet.
  • Features a securely closing hinged lid with a rust-resistant, nickel-plated clasp.
  • Stackable cases include feet for maximum stability.
Specifications
TypeSlide Storage Box
Slide Capacity100
Closure TypeHinged with Clasp
ColorWhite
Device Holds100 Microscope Slides
Dimensions (L x W x H)208 x 175 x 34 mm
For Use With (Equipment)Standard 25 x 75 mm or 1 x 3 in. microscope slides
Height (English)1.33 in.
Height (Metric)34 mm
IncludesSlide Inventory Sheet
Length (English)8.18 in.
Length (Metric)208 mm
MaterialABS
ShapeRectangular
StackableYes
Width (English)6.88 in.
Width (Metric)175 mm
Lining MaterialFoam
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link