5-Place Slide Mailer, End Opening
5-Place Slide Mailer, End Opening
Actual product may vary
Fisherbrand
5-Place Slide Mailer, End Opening
Ship or store slides safely. Fisherbrand™ Five-Slide Mailer is constructed of extra-strong, clear polypropylene with inner channels that keep the slides from touching one another. Snap-top lid closes securely to keep slides safe. Disposable and convenient.
Have Questions?
Catalog number FKS11719885
Price (SGD)
47.00
PK
Add to cart
Price (SGD)
47.00
PK
Add to cart
Ship or store slides safely. Fisherbrand™ Five-Slide Mailer is constructed of extra-strong, clear polypropylene with inner channels that keep the slides from touching one another. Snap-top lid closes securely to keep slides safe. Disposable and convenient.
  • Mailer holds five standard slides (25 x 75 mm and 1 x 3 in)
  • Inside wall features slotted channels to keep slides separated and to make removal easier
  • Lid closes securely to keep slides safe
  • Disposable
Specifications
TypeSlide Mailer
Slide Capacity5
Closure TypeSnap-top
ColorNatural
Dimensions (L x W x H)43 x 24 x 88 mm
For Use With (Equipment)Standard Slides
MaterialPolypropylene
Quantity25 Pk.
Unit SizePK