Check Order Status
Square HDPE Graduated 250 mL Bottles
Actual product may vary
Fisherbrand
Square HDPE Graduated 250 mL Bottles
Provides excellent storage for liquids and solids in the laboratory. Gosselin™ Square HDPE Graduated 250mL Bottles come with either a single closure, double closure, or leak-proof screw cap.
Have Questions?
Catalog Number
Closure Type
Closure Material
Certifications/Compliance
Closure Color
FKS11719592
Double Screw Cap
HDPE
EC N°1935/2004
Blue
Catalog number
FKS11719592
Price (SGD)
833.00
PK
Add to cart
Closure Type:
Double Screw Cap
Closure Material:
HDPE
Certifications/Compliance:
EC N°1935/2004
Closure Color:
Blue
Price (SGD)
833.00
PK
Add to cart
Break resistant
Molded graduations
Large mouth, wide neck
Translucent HDPE plastic
Aseptic or sterile by irradiation
Specifications
Capacity (Metric)
250 mL
Color
Natural
Description
Square
Diameter (Metric) Inner Neck
37 mm
Material
HDPE
Length (Metric)
57mm
Sterility
Non-sterile
Width (Metric)
57mm
Closure Color
Blue
Certifications/Compliance
EC N°1935/2004
Closure Included
Yes
Height (Metric)
109 mm
Closure Liner
Tresylene
Closure Material
HDPE
Closure Type
Double Screw Cap