Polypropylene Buckets with Handle
Polypropylene Buckets with Handle
Actual product may vary
Fisherbrand
Polypropylene Buckets with Handle
Excellent option for general storage in the laboratory. Gosselin™ Polypropylene Buckets with Handle are manufactured from white-colored polypropylene plastic.
Have Questions?
Catalog NumberCapacity (Metric)
FKS117195353800 mL
Catalog number FKS11719535
Price (SGD)
1,025.00
PK
Add to cart
Capacity (Metric):
3800 mL
Price (SGD)
1,025.00
PK
Add to cart
  • Nonsterile
  • Convenient carrying handle
  • Closure with tamper-evident seal
Specifications
Product TypeBucket
Capacity (Metric)3800 mL
MaterialPolypropylene
AutoclavableAutoclavable
Certifications/ComplianceEC N°1935/2004
ColorWhite
Diameter (Metric) Bottom167 mm
Diameter (Metric) Outer184 mm
Diameter (Metric)Top200 mm
Height (Metric)177 mm
IncludesSnap cap, Handles and Lids
Quantity75 Pk.
Unit SizePK