Clear Boston Round Bottles with Black Phenolic PolyCone Cap
Clear Boston Round Bottles with Black Phenolic PolyCone Cap
Actual product may vary
Fisherbrand
Clear Boston Round Bottles with Black Phenolic PolyCone Cap
General use bottles. Fisherbrand™ Clear Boston Round Bottles with Black Phenolic PolyCone Cap are ideal when content integrity is important.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeQuantity
FKS1171872960 mL20-400 mm24/Cs.
FKS1170872930 mL20-400 mm48/Cs.
Catalog number FKS11718729
Price (SGD)
-
Capacity (Metric):
60 mL
Closure Size:
20-400 mm
Quantity:
24/Cs.
Clear Glass Boston Round Bottles are excellent general use bottles ideal for when content integrity and sample or product visibility is important. Boston Round bottles feature a narrow screw neck opening, curved shoulders, and a round body. Glass offers minimal chemical interference with contents; clear (flint) glass is appropriate when visibility is important but protection from UV rays is not necessary. These environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage.
  • Features narrow screw neck opening, curved shoulders, and a round body
  • Glass offers minimal chemical interference with contents
Specifications
Capacity (Metric)60 mL
Capacity (English)2 oz.
ColorClear
MaterialGlass
Length (Metric)95mm
Quantity24/Cs.
Closure ColorBlack
Liner MaterialPolyCone
Neck Finish20-400
Closure IncludedYes
MouthNarrow
ShapeBoston Round
Height (Metric)94 mm
Product TypeBoston Round Bottle
Closure LinerPolyethylene
Closure MaterialPhenolic
Closure Size20-400 mm
Diameter (Metric)39 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePK