Gosselin™ Contact Dish
Gosselin™ Contact Dish
Actual product may vary
Fisherbrand
Gosselin™ Contact Dish
Optimized for automation--Gosselin™ Petri Dishes are manufactured from crystal polystyrene
Have Questions?
Catalog NumberSterility
FKS11718573Aseptic
Catalog number FKS11718573
Price (SGD)
632.00
PK
Add to cart
Sterility:
Aseptic
Price (SGD)
632.00
PK
Add to cart
  • 65 x 14.5 mm (D x H)
  • 0/3/4 vents
  • Crystal Polystyrene (PS)
  • Optimal flatness
  • Grip lid
  • Molded grid 10 x 10 mm
Specifications
Certifications/ComplianceEC N°1935/2004, ISO 11137, Aseptic
ColorNatural
MaterialPolystyrene
ShapeRound
SterilityAseptic
TypeContact Petri Plate
Well Count1
For Use With (Application)For use in clinical, environment, industries, microbiology
Quantity720 Carton
Height (Metric)14.5 mm
Diameter (Metric)65 mm
Unit SizePK