SHM-200 Series Stirring/Heating Mantle
SHM-200 Series Stirring/Heating Mantle
Actual product may vary
Fisherbrand
SHM-200 Series Stirring/Heating Mantle
The SHM-200 Series Stirring/Heating Mantle provides powerful stirring speeds up to 520 rpm and heating up to 450°C.
Have Questions?
Catalog NumberCapacityVoltage
FKS11718382500 mL (17 oz.)230 V
Catalog number FKS11718382
Price (SGD)
3,699.00
EA
Add to cart
Capacity:
500 mL (17 oz.)
Voltage:
230 V
Price (SGD)
3,699.00
EA
Add to cart
  • Bidirectional stirring with stir bar auto capture function.
  • Thermal insulating cartridge provides maximum heat transfer.
  • Chemical resistant polypropylene housing is ''safe to the touch.''
  • Stir your sample while heating up to 450°C (842°F).
  • These durable stirring/heating mantles feature a bidirectional magnetic stirrer with speeds up to 520 rpm.
  • An ''auto-recapture'' function reactivates stirring if it is interrupted or lost for any reason.
  • A built-in power controller with dial markings from 1 to 10 offer great repeatability.
  • Two colored lights indicate ''power on'' and ''heat on.''
  • The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
  • Unique ventilation slots and ceramic fiber insulation keep the chemical-resistant polypropylene exterior cool even while in use.
  • A safety earth (ground) screen and double fuses provide added safety.
Specifications
Certifications/ComplianceCE
Capacity500 mL (17 oz.)
Capacity (Metric)500 mL
Depth (English)12.25 in.
Depth (Metric)31.1 cm
For Use WithFlask (500 mL)
Height (English)5.94 in.
Height (Metric)15.1 cm
Voltage230 V
Wattage220 W
Weight (Metric) Shipping2.75 kg
Weight (English) Shipping97 oz.
Width (English)9.37 in.
Width (Metric)23.8 cm
Product TypeStirring Heating Mantle
Temperature (Metric)Ambient to 450°C
Amperage0.9 A
Capacity (English)17 oz.
For Use With (Application)Stirring and heating solutions
Unit SizeEA