Split Tube Handheld UV Lamp
Split Tube Handheld UV Lamp
Actual product may vary
Fisherbrand
Split Tube Handheld UV Lamp
Mask the SW or LW portion of the bulb using the snap-on wavelength selector. UVP Ultra Violet Product™ Split Tube Handheld UV Lamp utilizes a single split-tube for both shortwave and longwave UV light emissions.
Have Questions?
Catalog number FKS11718261
Price (SGD)
1,983.00
EA
Add to cart
Price (SGD)
1,983.00
EA
Add to cart
  • Lightweight, portable and AC powered
  • 6 watt portable lamp
  • Wavelengths: 365nm longwave and 254nm shortwave.
  • One split tube.
  • Durable plastic construction
  • Lamp cord: 2.4m
  • Dimensions: 378 x 81 x 64mm
  • Weight: 0.9kg
  • Use with J-129 stand for hands-free operation.
Specifications
Voltage230 V
Wattage6 W
Wavelength365/254 nm
Dimensions (L x W x H)378 x 81 x 64 mm
Electrical Requirements6 W, 230 V, 50/60 Hz
Hertz50/60 Hz
IncludesOne multiband tube and snap-on wavelength selector
Lighting Intensity1290/950 μW/cm2
MaterialPlastic
ModelUVGL-55
Packaging Quantity1 Ea.
Product TypePortable UV Lamp
Unit SizeEA