Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Glass Stopper
Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Glass Stopper
Actual product may vary
Fisher Catalogue
Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Glass Stopper
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Class A Volumetric Flask with Glass Stopper is available in both narrow neck and wide neck options in a variety of sizes. DE-M marking on neck provides quick and accurate volume check.
Have Questions?
Catalog NumberCapacity (Metric)Closure SizeNeck Style
FKS1171814810 mL10/19Wide
Catalog number FKS11718148
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
10 mL
Closure Size:
10/19
Neck Style:
Wide
  • Class A Volumetric Flask
  • Constructed of Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards
  • DAkkS calibration certificate available upon request
  • Calibrated to contain (TC, In)
  • Narrow neck and wide neck options available in a variety of sizes
  • DE-M marking provides quick and accurate volume check
  • Glass stopper included
Specifications
CalibrationCalibrated To Contain
Certifications/ComplianceDIN EN ISO 1042
Closure MaterialGlass
ColorClear
DescriptionDE-M Marking
Capacity (Metric)10 mL
MaterialBorosilicate Glass
Limit of Error±0.04 mL
Neck StyleWide
No. per Case2 Pack
GraduatedYes
ClassA
Closure Size10/19
Unit SizePK