1x1 image pixel for data collection
DURAN™ Reagent Bottle, Narrow Neck, Clear
DURAN™ Reagent Bottle, Narrow Neck, Clear
Actual product may vary
Fisherbrand
DURAN™ Reagent Bottle, Narrow Neck, Clear
Classic narrow-neck, glass-stoppered reagent bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS117168581000 mL
FKS11706858500 mL
Catalog number FKS11716858
Price (SGD)
2,143.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Capacity (Metric):
1000 mL
Price (SGD)
2,143.00
PK
Add to cart
The classical stoppered reagent bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a narrow conical ground joint neck and comply with ISO 4796-2. The narrow neck is suitable for liquids and the angle of the shoulders ensures that the contents can be smoothly poured. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel.
  • All borosilicate glass components and therefore suitable for storage of aggressive materials
  • Tight closure due to standard ground joint
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic print
Specifications
Capacity (Metric)1000 mL
DescriptionNeck with standard ground joint and glass flat-headed stopper
For Use With (Application)Storage of air sensitive reagents, pharmaceuticals, and chemicals; Storage of hygroscopic liquids and pharmaceuticals; Transport and shipping of samples; Storage of hygroscopic liquids and pharmaceuticals
MaterialBorosilicate Glass (3.3)
Quantity10/Pk.
AutoclavableAutoclavable
Diameter (Metric)107 mm
Unit SizePK
Documents & Downloads
Certificates
Share catalog number, name or link