Wash Bottles with Adjustable Sprayer
Wash Bottles with Adjustable Sprayer
Actual product may vary
Fisherbrand
Wash Bottles with Adjustable Sprayer
Handle hot/cold, aqueous/solvent-based solutions
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11714356240 mL
CTL03-405-A240 mL
Catalog number FKS11714356
Price (SGD)
95.00
PK
Add to cart
Capacity (Metric):
240 mL
Price (SGD)
95.00
PK
Add to cart
Handle hot/cold, aqueous/solvent-based solutions using Fisherbrand™ Wash Bottles with Adjustable Sprayer.
Fully adjustable spray pattern: 'fine mist' for TLC plates; 'needle' for rinsing glassware; 'big stream' for powerful jet needed to wash filters.
  • Bottle is made of high-density polyethylene
  • Sprayer is polypropylene
  • Trigger allows one-handed, controlled delivery with light pressure
  • Engraved markings show how much fluid is left
  • Squat shape takes up little bench space and ensures a low center of gravity
Specifications
Capacity8 oz., 240mL
Capacity (Metric)240 mL
Capacity (English)8 oz.
ColorWhite
IncludesPolypropylene adjustable sprayer
MaterialHDPE
Quantity3/Pk.
SterilityNon-sterile
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
GraduatedNo
MouthThreaded
ShapeSquat
Height (Metric)10.16 cm
Product TypeSpray Bottle
Width (English)8.25 in.
Length (English)10.5 in.
Height (English)4 in.
Closure MaterialPolypropylene
Closure SizeFor 240 mL Bottle
Diameter (Metric)20.955 cm
Closure TypeThreaded
Unit SizePK