Polypropylene Microscope Slide Mailers
Polypropylene Microscope Slide Mailers
Actual product may vary
Fisherbrand
Polypropylene Microscope Slide Mailers
Keep slides safe and free from damage. Polypropylene Microscope Slide Mailers include channel slots to prevent breakage.
Have Questions?
Catalog NumberWidth (Metric) Exterior
FKS1171431619 mm
Catalog number FKS11714316
Price (SGD)
48.00
PK
Add to cart
Width (Metric) Exterior:
19 mm
Price (SGD)
48.00
PK
Add to cart
  • Polypropylene containers hold five standard slides (25 x 75 mm and 1 x 3 in.)
  • Inside wall features slotted channels to keep slides separated and to make removal easier
  • Lid closes securely to keep slides safe.
  • Pack of 25
  • Side-opening mailers measure 81 x 19 x 31 mm
  • Disposable
Specifications
Quantity25 Pk.
Width (Metric) Exterior19 mm
Length (English) Exterior3.2 in.
Width (English) Exterior0.7 in.
IncludesSlotted channels
Length (Metric) Exterior81 mm
ColorNatural
Height (English) Exterior1.2 in.
TypeSlide Mailer
Height (Metric) Exterior31 mm
Dimensions (LxWxH)81 x 19 x 31 mm
Capacity5 Slides