DeckWorks™ Hinged Rack Barrier Tips
DeckWorks™ Hinged Rack Barrier Tips
Actual product may vary
Fisherbrand
DeckWorks™ Hinged Rack Barrier Tips
Features an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures.
Have Questions?
Catalog NumberVolume (Metric)
FKS11709325100 to 1000 μL
Catalog number FKS11709325
Price (SGD)
639.00
PK
Add to cart
Volume (Metric):
100 to 1000 μL
Price (SGD)
639.00
PK
Add to cart
  • The hinged rack design allows easy rack stacking
  • Racks can be reused
  • Simply remove the tip insert in the rack and replace with the Corning tip inserts for hinged racks
  • Allow maximize storage space and help reduce the production and waste of plastic materials
  • Hinged racks within this system are manufactured from recycled polypropylene
  • Used for quantitative assays in which sample binding can significantly alter results
  • These sterile tips will not trap liquids or inhibit PCR in the event of over-pipetting
Specifications
Volume (Metric)100 to 1000 μL
FilteredFiltered
SterilitySterile
ColorNatural
Clean ClaimsNonpyrogenic, Human DNA-free, RNase-/DNase-free, Sterility Assurance Level 10-3
For Use With (Application)Used for quantitative assays in which sample binding can significantly alter results
FormatHinged Rack
Tip StyleRound
Packaging96 Tips/Rack, 8 Racks/Pack
GraduatedYes
Product LineDeckWorks
MaterialHDPE
Unit SizePK