Aura™ Particulate Respirators 9300+ Series
Aura™ Particulate Respirators 9300+ Series
Actual product may vary
Aura™ Particulate Respirators 9300+ Series
Aura™ Particulate Respirators 9300+ Series
Aura™ Particulate Respirators 9300+ Series
Aura™ Particulate Respirators 9300+ Series
Aura™ Particulate Respirators 9300+ Series
Fisherbrand
Aura™ Particulate Respirators 9300+ Series
Particulate respirators have an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media. Available in FFP1, FFP2, and FFP3 protection levels with or without valves, Innovative chin tab for easy adjustments and an embossed, sculpted upper panel helps reduce eyewear fogging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFilter TypeWith Exhalation ValveAssigned Protection Factor (APF)Model
FKS11707584FFP3Yes20PF9332
9330FFP3No209330
Catalog number FKS11707584
Price (SGD)
178.00
PK
Add to cart
Filter Type:
FFP3
With Exhalation Valve:
Yes
Assigned Protection Factor (APF):
20
Model:
PF9332
Price (SGD)
178.00
PK
Add to cart
Particulate respirators have an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media. Available in FFP1, FFP2, and FFP3 protection levels with or without valves, Innovative chin tab for easy adjustments and an embossed, sculpted upper panel helps reduce eyewear fogging.
  • Provide comfortable protection against dusts and mists in a wide variety of industrial applications and other hazardous environments.
  • Offers FFP1, FFP2 and FFP3 protection.
  • 3-panel design allows for greater facial movement.
  • Sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging and improves eyewear compatibility.
  • Low resistance filter technology promotes easier breathing and the innovative chin tab allows simple adjustments.
  • Choose without valve style or with valve styles that feature 3M™ Cool Flow™ to reduce heat build-up and help provide comfortable protection in hot and humid conditions.
  • Individually packed in hygienic wrapping to prevent contamination before use.
  • Braided, colour-coded headbands help you identify the protection level offered: yellow for FFP1 (APF 4), blue for FFP2 (APF 10) and red for FFP3 (APF 20).
  • Foldable design makes them easy to store when not in use.
  • Respirators work well with 3M eyewear and hearing protection products.
Specifications
Assigned Protection Factor (APF)20
Certifications/ComplianceCE Approved to EN 149:2001A1:2009
Certifications/Compliance (Europe)EN 149:2001 + A1:2009
Certifications (EN)EN 149:2001 + A1:2009
ColorWhite
Design TypeFlat-fold
DisposableLimited-use
Filter TypeFFP3
MaterialPolypropylene
Product TypeGeneral Purpose Particulate Respirator
SizeOne Size
With Exhalation ValveYes
Filter EfficiencyFFP3
Filtration Layer MaterialPolypropylene
For Use With (Application)Cement Dust
Latex FreeYes
Material HeadstrapPolyisoprene
ModelPF9332
Nose Piece MaterialAluminum, Polyurethene
Nosepiece TypeSculpted
Packaging TypeIndividually Packaged
Resistant ToDusts and mists found in a wide variety of industrial applications and other work situations
Shelf Life5 Years From Date of Manufacture
SterilityNon-sterile
Style3-panel
Strap Attachment TypeColour-coded Headband, Red
FDA Cleared Surgical MaskNo
Unit SizePK