DURAN™ Silicone Septum Seal, Peroxide cured silicone, Septum for Piercing, Use with Open Topped Screw Cap
DURAN™ Silicone Septum Seal, Peroxide cured silicone, Septum for Piercing, Use with Open Topped Screw Cap
Actual product may vary
DURAN™ Silicone Septum Seal, Peroxide cured silicone, Septum for Piercing, Use with Open Topped Screw Cap
DURAN™ Silicone Septum Seal, Peroxide cured silicone, Septum for Piercing, Use with Open Topped Screw Cap
Fisherbrand
DURAN™ Silicone Septum Seal, Peroxide cured silicone, Septum for Piercing, Use with Open Topped Screw Cap
Silicone Septum Seal for piercing
Have Questions?
Catalog NumberThread Size (Metric)
FKS11707198GL14
Catalog number FKS11707198
Price (SGD)
369.00
PK
Add to cart
Thread Size (Metric):
GL14
Price (SGD)
369.00
PK
Add to cart
Suitable for PBT open topped screw cap with central aperture Heat resistance: 130°C (steam) and 200°C (dry heat). Silicon is peroxide catalysed.
  • Compatible with open topped GL screw caps with central aperture
  • Compatible with GL bottles
  • Peroxide catalysed silicone
Specifications
For Use With (Application)Liquid transfer from HPLC solvent bottles; Aseptic reagent delivery or culture sampling using a syringe and needle; Reservoir cap for supplying laboratory instruments
MaterialSilicone
Quantity100/Pk.
Thickness (Metric)2 mm
Thread Size (Metric)GL14
AutoclavableAutoclavable
Diameter (Metric)12 mm
Unit SizePK