Check Order Status
Economical Hemacytometer with Cover Slips
Actual product may vary
Fisherbrand
Economical Hemacytometer with Cover Slips
Economical Hemacytometer with Cover Slips is specially designed for counting erythrocytes and leukocytes.
Have Questions?
Catalog number
FKS11704939
Price (SGD)
1,715.00
EA
Add to cart
Price (SGD)
1,715.00
EA
Add to cart
Two special counting chamber glass coverslips.
Light transmission between 29 and 32%.
This hemacytometer features high light transmission and brightly illuminated line.
Double rulings are visible through a wide range of illumination.
Neubauer rulings are 0.1 mm below cover glass.
The H-shaped moat offers two counting areas.
Fused rhodium on glass provides distinct lines.
Two glass coverslips are included.
Specifications
Product Type
Hemacytometer
Grid Type
Neubauer
Quantity
1 Ea.
For Use With (Application)
For counting erythrocytes and leukocytes
Includes
Cover Slips