Magnifiers
Magnifiers
Actual product may vary
Fisherbrand
Magnifiers
Applied for laboratory use and other precision scientific work
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMagnificationDiameter (English) Outer
FKS117043963x1.75 in.
CTL12-071-6B3x1.75 in.
Catalog number FKS11704396
Price (SGD)
101.00
EA
Add to cart
Magnification:
3x
Diameter (English) Outer:
1.75 in.
Price (SGD)
101.00
EA
Add to cart
Fisher Scientific™ Magnifier Lamp clamps vertically or horizontally to top or side of any edge. Applied for laboratory use and other precision scientific work.
Large Magnifier
  • Illuminated 2X magnification with 4X spot
  • Window is 2.75 x 2.75 in. (7 x 7 cm)
  • Overall dimensions: 8.5L x 3W x 1 in.H (21.6 x 7.6 x 2.5 cm)
Medium Magnifier
  • Illuminated dual 3X and 10X magnifiers
  • Window is 1.75 x 1.75 in. (4.4 x 4.4 cm)
  • Overall dimensions: 6.5L x 2W x 0.75 in.H (16.5 x 5.1 x 1.9 cm)
Small Magnifier
  • 8X Magnification
  • Adjustable focusing
  • Window is 1 in. (2.5 cm) dia.
Specifications
Diameter (English) Outer1.75 in.
Diameter (Metric) Outer4.44 cm
Dimensions (L x W x H)6.5 x 2 x 0.75 in. (16.51 x 5 x 1.91cm)
For Use With (Application)To view surface details and illuminate uniformly over the field of observation
Height (English)0.75 in.
Height (Metric)1.91 cm
IncludesAA replaceable battery
Length (English)6.5 in.
Length (Metric)16.51 cm
LightingLED
Weight (English)4 oz.
Weight (Metric)113.39 g
Width (English)2 in.
Width (Metric)5 cm
Diameter (Englis) Lens1.75 in.
Diameter (Metric) Lens4.44 cm
Magnification3x
MaterialPlastic
Product TypeMagnifier, Illuminated
Unit SizeEA