GasPak™ Supplies
GasPak™ Supplies
Actual product may vary
Fisherbrand
GasPak™ Supplies
Used with the GasPak™ EZ Gas Generating Systems. BD™ GasPak™ Supplies are used with these complete, integrated systems for rapid, waterless and catalyst-free gas generation. Can be used to support an anaerobic, microaerophilic or CO2 enriched environment.
Have Questions?
Catalog NumberTypeQuantity
FKS11703473Dry Anaerobic Indicator Strip100 Pk.
Catalog number FKS11703473
Price (SGD)
758.00
PK
Add to cart
Type:
Dry Anaerobic Indicator Strip
Quantity:
100 Pk.
Price (SGD)
758.00
PK
Add to cart
GasPak™ EZ Gas Generating Systems are complete, integrated systems that rapidly produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
Accessories include:
  • CO2 Indicator Strips
  • Dry Anaerobic Indicator Strips
  • Tube Holder
  • Catalyst Replacement Charges
  • Catalyst Reaction Chamber
Specifications
TypeDry Anaerobic Indicator Strip
DescriptionStrip changes from blue to colorless in the absence of oxygen.
FormatStrip
For Use With (Application)Produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria
For Use With (Equipment)BD GasPak™
Quantity100 Pk.
Unit SizePK
Contents & Storage
15°C to 30°C