DURAN™ High Temperature Screw Cap, PTFE-Coated, Peroxide cured silicone, Red PBT
DURAN™ High Temperature Screw Cap, PTFE-Coated, Peroxide cured silicone, Red PBT
Actual product may vary
DURAN™ High Temperature Screw Cap, PTFE-Coated, Peroxide cured silicone, Red PBT
DURAN™ High Temperature Screw Cap, PTFE-Coated, Peroxide cured silicone, Red PBT
Fisherbrand
DURAN™ High Temperature Screw Cap, PTFE-Coated, Peroxide cured silicone, Red PBT
Screw cap with cap liner sealing and high temperature and chemical resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThread Size (Metric)
FKS11702342GL 32
FKS10222161GL 45
Catalog number FKS11702342
Price (SGD)
221.00
PK
Add to cart
Thread Size (Metric):
GL 32
Price (SGD)
221.00
PK
Add to cart
The red polybutylene terephthalate (PBT) GL cap and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. Screw cap seal has a PTFE faced cap liner for efficient sealing. ETFE pouring ring ensures drip free pouring.
  • Excellent chemical resistance due to PTFE coating
  • Cap liner forms a tight seal with bottle Peroxide catalysed silicone
  • Peroxide catalysed silicone
  • Suitable for PBT screw caps and tamper-evident screw caps
  • Fully compliant with current international food contact regulations
Specifications
ColorRed
DescriptionScrew cap
For Use With (Application)Autoclaving of media and reagents; Preparation of media and reagents; Suitable for dry heat sterilization; Pressure applications; Storage of corrosive reagents; Chemical and thermal reactions; Long term / archival storage of samples, media and reagents; Sampling; Transport and shipping
MaterialPBT, ETFE, Fiberglass
Quantity10/Pk.
Screw MaterialPolybutylene Terephthalate (PBT) with glass fiber
Thread Size (Metric)GL 32
Liner MaterialPTFE-coated Silicone
AutoclavableNot Autoclavable
Closure LinerPTFE/Silicone
Closure MaterialPBT with Glass Fiber
Diameter (Metric)41 mm
Unit SizePK