Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Polypropylene Stopper
Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Polypropylene Stopper
Actual product may vary
Fisherbrand
Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Polypropylene Stopper
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flask with Polypropylene Stopper is available with narrow neck or wide neck in a variety of sizes. DE-M marking on the neck provides quick and accurate volume check. Polypropylene stopper included.
Have Questions?
Catalog NumberCapacity (Metric)Neck Style
FKS1169319910 mLWide
Catalog number FKS11693199
Price (SGD)
105.00
PK
Add to cart
Capacity (Metric):
10 mL
Neck Style:
Wide
Price (SGD)
105.00
PK
Add to cart
  • Class A Volumetric Flask
  • Constructed of Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards
  • DAkkS calibration certificate available upon request
  • Calibrated to contain (TC, In)
  • Narrow neck and wide neck options available in a variety of sizes
  • DE-M marking provides quick and accurate volume check
  • Polypropylene stopper included
Specifications
CalibrationCalibrated To Contain
Certifications/ComplianceDIN EN ISO 1042
Closure MaterialPolypropylene
ColorClear
DescriptionDE-M Marking
Capacity (Metric)10 mL
MaterialBorosilicate Glass
Limit of Error±0.04 mL
Neck StyleWide
No. per Case2 Pack
GraduatedYes
ClassA
Closure TypeStopper
Closure Size10/19
Unit SizePK