Orion™ Versa Star Conductivity Measurement Module
Orion™ Versa Star Conductivity Measurement Module
Actual product may vary
Orion™ Versa Star Conductivity Measurement Module
Orion™ Versa Star Conductivity Measurement Module
Orion™ Versa Star Conductivity Measurement Module
Fisherbrand
Orion™ Versa Star Conductivity Measurement Module
Add conductivity, TDS, salinity, resistivity and temperature measurement capabilities to your Versa Star meter using the Thermo Scientific™ Orion™ Versa Star Conductivity Module
Have Questions?
Catalog number FKS11691509
Price (SGD)
4,644.00
EA
Add to cart
Price (SGD)
4,644.00
EA
Add to cart
Add conductivity, total dissolved solids (TDS), salinity, resistivity and temperature measurement capabilities to your Thermo Scientific™ Orion™ Versa Star meter using the Thermo Scientific™ Orion™ Versa Star Conductivity Measurement Module. Each Orion™ Versa Star meter has four channels that can be customized with interchangeable measurement modules. The Versa Star conductivity module offers reliable, fast results with auto-ranging conductivity values and linear, non-linear pure water or EP temperature compensation with 5, 10, 15, 20 or 25 °C reference temperatures.
Select from five Thermo Scientific Orion Versa Star measurement modules to customize the measurement capabilities of your Orion Versa Star meter. Measurement modules are interchangeable and can be added or removed from each Versa Star meter's four channels at any time.
Orion Versa Star Conductivity Measurement Module:
  • Measure conductivity, TDS, salinity or resistivity with temperature
  • Perform up to a six point conductivity calibration with calibration editing option that allows individual points to be fixed without a full recalibration
  • Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate
  • Use linear, non-linear, EP (USP) or off conductivity temperature compensation with 5, 10, 15, 20 or 25°C reference temperatures
  • Customize measurements to show calibration history or simplify to show an extra large conductivity reading
  • Module offers a 3-year replacement warranty
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy (Conductivity)0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
Accuracy (Resistivity)0.5% of reading ±1 digit
Accuracy (TDS)0.5% of reading ±1 digit
Accuracy (Temperature)±0.1°C
Accuracy (Salinity)±0.1ppt
Alarm OutputSet point alarm, high/low limit alarm, calibration due alarm
CalibrationConductivity with calibration editing option and temperature
Cell Constant0.001 to 199.9
Calibration PointsUp to 6
Compatible Probes2-cell or 4-cell conductivity sensors with built-in temperature
DescriptionOrion VERSA STAR Conductivity/TDS/Resistivity/Salinity/Temperature Module
TypeConductivity Measurement Module
Dimensions (L x W x H)12 x 3 x 9.5 cm (4.7 x 1.2 x 3.8 in.)
Inputs8 pin MiniDIN (conductivity sensor with built-in temperature)
IncludesVersa Star conductivity module and printed quick start guide
IP RatingIP-54
Measurement ModesConductivity, TDS, salinity or resistivity with temperature
Methods10 per channel with password protection
ModelOrion Versa Star Conductivity/TDS/Salinity/Resistivity/Temperature Measurement Module
Probe TypeConductivity sensor (purchase separately)
Range (Conductivity)0.001 µS/cm to 3000 mS/cm
Range (Resistivity)2 ohm to 100.0 meg-ohm
Range (Salinity)0.06 to 80.00 psu, 0.05 to 42.00 ppt
Range (TDS)0.001 ppm to 200.0 ppt
Resolution (Conductivity)0.001 μS minimum, auto ranging up to 4 significant digits
Relative Humidity Range5 to 85 %, non-condensing
Resolution (Resistivity)1 ohm or 0.1 meg-ohm, auto ranging
Resolution (Salinity)0.01 psu or 0.01 ppt minimum, auto ranging
Resolution (Temperature)0.1°F/°C
Resolution (TDS)0.001 ppm minimum, auto ranging up to 4 significant digits
Salinity Measurement ModesPractical salinity (psu) or natural sea water (ppt)
StabilitySelectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
Temperature CompensationLinear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off
TDS Factor RangeLinear (0.02 to 9.99)
Temperature (English) Range41°F to 113°F
Temperature (Metric) Range-5°C to 105deg;C
Temperature Probe Calibration1 point temperature offset calibration
Temperature (Metric) Operating5°C to 45°C
Temperature (Metric) Reference5, 10, 15, 20, 25°C
Temperature SelectionManual or automatic with ATC temperature probe
Weight (English)0.5 lb.
Warranty3 year meter replacement
Weight (Metric)0.2 kg
Unit SizeEA