Orion Star™ A222 Conductivity Portable Meter
Orion Star™ A222 Conductivity Portable Meter
Actual product may vary
Fisherbrand
Orion Star™ A222 Conductivity Portable Meter
Reliably measure conductivity, TDS, salinity, resistivity and temperature with this conductivity portable meter for advanced field analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProbe Type
FKS116614694-cell Conductivity Sensor
FKS11651469Not included
FIS13-645-5314-cell Conductivity Sensor
Catalog number FKS11661469
Price (SGD)
5,917.00
EA
Add to cart
Probe Type:
4-cell Conductivity Sensor
Price (SGD)
5,917.00
EA
Add to cart
Record accurate and reliable conductivity, total dissolved solids (TDS), salinity, resistivity and temperature measurements with the Thermo Scientific™ Orion Star™ A222 Conductivity Portable Meter. This instrument is ideal for a wide range of applications and advanced conductivity analysis in the field. Obtain temperature compensated readings using linear, non-linear or EP options with 15°C, 20°C or 25°C reference temperatures. Log up to 1000 data point sets with time/date stamp and transfer via USB or RS232 to a printer or computer. Waterproof meter ensures durable, rugged performance with an IP67-rated housing.
Don't leave your measurements to chance when analyzing samples outside the lab. Orion Star A222 conductivity portable meters are designed to offer the reliability you need in a user-friendly package for your dedicated conductivity testing needs. This waterproof portable meter displays critical data with your measurement including temperature, date and time, read type, measurement stability, active calibration info and power source status.
  • Measure conductivity, TDS, salinity or resistivity with temperature
  • Perform up to a five point conductivity calibration
  • Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate
  • Use linear, non-linear or off conductivity temperature compensation with 15°C, 20°C or 25°C reference temperatures
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes – Auto-Read holds a stable reading and Continuous shows changing readings with hold option
  • Data log collects and date/time stamps up to 1000 measurement sets
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications and 3-year replacement warranty
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy (Conductivity)0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
DescriptionOrion Star A222 conductivity/TDS/salinity/resistivity portable meter kit with DuraProbe 4-cell conductivity sensor, solutions, meter armor and field case
OutputsUSB, RS232
IncludesStar A222 Conductivity Meter with Batteries; 013010MD DuraProbe Epoxy 4-Probe (K=0.475) Conductivity Probe with 3 Meter Cable; 01100710 Conductivity 1413uS Standard Pouches; 911110 Rinse Solution Pouches; STARA-CS Star A Meter Hard Carrying Case; STARA-AR Protective Armor with Electrode Holder
Measurement ModesConductivity, TDS, Salinity, Resistivity with Temperature
Compatible Probes2-cell or 4-cell conductivity sensors with built-in temperature
Cell Constant0.001 to 199.9cm-1
CalibrationConductivity and temperature
Calibration Points (Conductivity)5
Range (Conductivity)0.001 μS/cm to 3000 mS/cm
Resolution (Conductivity)0.001 μS minimum, auto ranging up to 4 significant digits
Temperature CompensationLinear, nLFn (non-linear ultra pure water) or off
Temperature (Metric) Reference15, 20, 25°C
Range (Salinity)0.06 to 80.00 psu, 0.05 to 42.00 ppt
Resolution (Salinity)0.01 psu or 0.01 ppt minimum, auto ranging
Accuracy (Salinity)0.5% of reading ±1 digit
Salinity Measurement ModesPractical salinity (psu)
Resolution (TDS)0.001 ppm minimum, auto ranging up to 4 significant digits
Accuracy (TDS)0.5% of reading ±1 digit
TDS Factor RangeLinear (0.02 to 9.99)
Temperature Range (Metric)-5°C to +105°C
Resolution (Temperature)0.1°C (0.1°F)
Accuracy (Temperature)±0.1°C
Range (Resistivity)2 ohm to 100.0 megohm
Temperature SelectionManual or automatic with ATC temperature probe
Temperature Probe Calibration1 point temperature offset calibration
Display TypeGraphic LCD with Backlight
Backlight OptionYes (Selectable)
MemoryNon-volatile memory preserves data log, calibration log and meter settings
Data Points1000 with Date and Time Stamp
Log Function TypeAutomatic with Auto-Read Measure Mode, Manual with Continuous Measure Mode
Data Management DetailsTransfer single, range or all data points to printer or computer
Alarm OutputCalibration due alarm (fixed 12 hour)
Inputs8 pin MiniDIN (conductivity sensor with built-in temperature)
Certifications/ComplianceCE, TUV 3-1, FCC Class A
IP RatingIP67
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
Dimensions (L x W x H)24 x 10 x 6.5 cm (9.5 x 3.9 x 2.6 in.)
Weight (Metric)0.5 kg
Weight (English)1.2 lb.
Warranty3 Year Meter
Probe Type4-cell Conductivity Sensor
Relative Humidity Range5 to 85%, non-condensing
ModelOrion Star A222 Conductivity/TDS/Salinity/Resistivity/Temperature Portable Meter
Resolution (Resistivity)1 ohm or 0.1 megohm, auto ranging
Power Supply4 AA batteries (included) or universal AC adapter
Methods10 per channel with password protection
StabilitySmart Stability and Smart Averaging
KeypadComprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
LanguagesEnglish, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Channels1
Accuracy (Resistivity)0.5% of reading ±1 digit
Battery Type4 x AA (included)
Battery Life800 hr.
Unit SizeEA