Orion Star™ A111 Benchtop pH Meter
Orion Star™ A111 Benchtop pH Meter
Actual product may vary
Fisherbrand
Orion Star™ A111 Benchtop pH Meter
Take simple pH, mV and temperature measurements with our benchtop pH meter with easy operation for basic lab testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
FKS11661399pH Meter
FIS13-645-501pH Meter
Catalog number FKS11661399
Price (SGD)
2,333.00
EA
Add to cart
Type:
pH Meter
Price (SGD)
2,333.00
EA
Add to cart
Take simple, routine pH, mV and temperature measurements with the Thermo Scientific™ Orion Star™ A111 Benchtop pH Meter. This meter is an easy-to-use and budget-friendly instrument for basic pH analysis in the lab. Perform up to a 3 point pH calibration with easy recall of calibration points and slope. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
Features:
  • Large, informative screen clearly shows the main information, including pH or mV reading, temperature, meter mode, calibration information and power source.
  • Thermo Scientific™ AUTO-READ™ locks in the stable reading on your screen and ready indicator alerts when readings are stable.
  • Meter offers up to 3 point pH calibration and automatic recognition of USA/NIST and DIN buffers.
  • Use can easily recall calibration slope data for procedure checks.
  • Nonvolatile memory holds up to 50 data points.
  • Included electrode arm and newly designed probe holder make it easier to maintain and place probes into samples.
  • Plug the meter into almost any power outlet with the included universal power adapter or use four AA batteries (sold separately) to run the meter on DC power.
  • The IP54-rated housing can handle splashes and can be wall-mounted if table space is a problem.
Includes:
Orion STAR: A111 Benchtop pH Meter, Part no. STARA1110, includes electrode arm with redesigned holder and universal power adapter
Orion STAR: A111 Benchtop pH Meter Kit, Part no. STARA1115, includes 9157BNUMD Orion Triode™ 3-in-1 pH/ATC probe; Solution kit (60mL each of pH 4, 7 and 10 buffers; storage solution and cleaning solution); electrode arm with redesigned holder and universal power adapter
Orion STAR: A111 Benchtop pH Meter Kit, Part no. STARA1116, includes GD9156BNWP Orion Green electrode; 927007MD Stainless Steel ATC Probe; electrode arm with redesigned holder and universal power adapter
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 or ±0.05%, whichever is greater
Accuracy (pH)±0.01
Data Management DetailsInternal Data Point Memory
DescriptionOrion Star A111 benchtop pH meter with stand
OutputsNone
IncludesStar A111 pH meter, electrode stand, 100-240V universal power adapter
Measurement ModespH, mV, Relative mV with Temperature
CalibrationpH, relative mV (RmV) and temperature
Range (pH)-2 to 16
Resolution (pH)0.01, 0.1
Range (mV)±1600.0
Resolution (mV)0.1mV
Calibration Points1 (relative mV)
Temperature Range (Metric)-5°C to +105°C
Temperature Range (English)22.0°F to 221.0°F
Resolution (Temperature)0.1°C, 0.1°F
Temperature SelectionManual or automatic with ATC temperature probe
Temperature Probe Calibration1 point temperature offset calibration
Display TypeLCD
MemoryNon-volatile memory preserves log and settings
Data Points50
Log Function TypeAutomatic with Auto-Read Measure Mode, Manual with Continuous Measure Mode
InputsBNC (pH or ORP electrode), 8 pin MiniDIN (ATC temperature probe)
Certifications/ComplianceCE, TUV 3-1, FCC Class A
IP RatingIP54
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
Dimensions (L x W x H)24 cm x 18 cm x 11 cm (9.5 in. x 7.1 in. x 4.3 in.)
Weight (Metric)0.9 kg
Weight (English)2 lb.
Warranty3 Year Meter Warranty
KeypadSimple with highlighted measure key, scroll and shortcut keys
ModelOrion Star A111 Benchtop pH/mV/RmV/Temperature Meter
Power SupplyUniversal AC adapter (included) or 4 AA batteries (sold separately)
Probe TypepH electrode, ORP electrode, ATC temperature probe (purchase separately)
LanguagesEnglish
Range (Relative mV)±1999.9mV
Relative Humidity Range5 to 85%, non-condensing
Isopotential Point7.00pH
StabilitySmart Stability and Smart Averaging
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Channels1
Battery Type4 x AA (sold separately)
TypepH Meter
Battery Life2000 hr.
Unit SizeEA