Provides stable stirring action. The plain economy PTFE magnetic stir bar work within a wide range of conditions, low prices from high-volume manufacturer.
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric)
FKS11597572
25 mm
8 mm
FKS11517582
50 mm
8 mm
FKS11507582
40 mm
8 mm
3 Options
Catalog number FKS11597572
Price (SGD)
39.00
PK
Add to cart
Length (Metric):
25 mm
Diameter (Metric):
8 mm
Price (SGD)
39.00
PK
Add to cart
Provides stable stirring action. The plain economy PTFE magnetic stir bar work within a wide range of conditions, low prices from high-volume manufacturer.
Stable, general-purpose magnetic stir bar
Smooth, round profile for a wide variety of applications
Autoclavable
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates