Traceable™ Three-Line Alarm Timer
Traceable™ Three-Line Alarm Timer
Actual product may vary
Traceable™ Three-Line Alarm Timer
Traceable™ Three-Line Alarm Timer
Fisherbrand
Traceable™ Three-Line Alarm Timer
Perfect unit to dedicate to a group of repetitive tests
Have Questions?
Catalog number FKS11597453
Price (SGD)
161.00
EA
Add to cart
Price (SGD)
161.00
EA
Add to cart
The Fisher Scientific™ Traceable™ Three-Line Alarm Timer is a user-friendly timer that displays three different countdown/alarm times simultaneously. Three unique channels may be started individually or concurrently and programmed to count down from 99 hours, 59 minutes, 59 seconds to 1 second. When zero is reached, an alarm sounds, TIME's UP is displayed, and the timer begins counting up. It's the perfect unit to dedicate to a group of repetitive tests.
  • Memories return the display to the previously programmed time
  • May also be used as three stopwatches or in any combination of up/down timing
  • Display has a time-of-day clock
  • Quartz-crystal timing accuracy is 0.01%
  • Digits are 3/8 in. high
  • Chemical-resistant ABS plastic case size: 2 x 3.25 x 0.5 in.
  • Weight: 3 oz.
Specifications
Product TypeHandheld/Benchtop Timer
Time Range99 hr. 59 min. 59 sec.
AlarmsAudible/Visible
Accuracy0.0001
Resolution1 sec.
DisplayLCD
Battery Type357 Button-Cell Silver Oxide Battery (Cat. No. 02-440-1C)
Height (English) Display0.37 in.
Height (Metric) Display0.93 cm
Weight (English)3 oz.
Weight (Metric)85.04 g
IncludesIndividually serial-numbered Traceable Certificate, magnetic back, easel stand, and 1.5-year silver-oxide battery
Certifications/Complianceto +assure accuracy, an individually serial-numbered Traceable™ Certificate is provided from our ISO/IEC 17025 calibration laboratory accredited by A2LA.  It includes traceability to +standards provided by the NIST (National Institute of Standard
Unit SizeEA