Glutathione Sepharose™ 4B Media
Glutathione Sepharose™ 4B Media
Actual product may vary
Glutathione Sepharose™ 4B Media
Glutathione Sepharose™ 4B Media
Fisherbrand
Glutathione Sepharose™ 4B Media
Use for high capacity purification of Glutathione S-Transferase (GST) tagged fusion proteins. Cytiva Life Sciences™ Glutathione Sepharose™ 4B Media requires only a single-step for purification.
Have Questions?
Catalog NumberQuantity
FKS1159493510 mL
Catalog number FKS11594935
Price (SGD)
-
Quantity:
10 mL
  • Binding capacity: >5mg horse liver GST/mL medium (flowrate dependent) Average particle size: 90μm
  • Recommended flowrate (H2O at room temp.): 75cm/hour
  • Stable in presence of all commonly used aqueous buffers
  • Stable in pH range 4 to 13
Specifications
MaterialGlutathione Sepharose 4 B
pH Range4 to 13
Particle Size90 μm
Quantity10 mL
Unit SizeEA