Stainless Steel Straight Tongs
Stainless Steel Straight Tongs
Actual product may vary
Fisherbrand
Stainless Steel Straight Tongs
Get maximum results using laboratory tongs in high temperature chemical reactions. Made of durable stainless steel, these Clifton™ Stainless Steel Straight Tongs ensure precise control of grasping, lifting, turning, and transferring heat-resistant and other materials in a laboratory environment.
Have Questions?
Catalog NumberLength (Metric)
FKS11593492150 mm
Catalog number FKS11593492
Price (SGD)
25.00
EA
Add to cart
Length (Metric):
150 mm
Price (SGD)
25.00
EA
Add to cart
  • Laboratory straight tongs
  • Stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Designed for corrosion resistance and durability ideal in laboratory routines
  • Laboratory metalware consumables
  • Finest in corrosion-resistant 304 stainless steel
  • Provides corrosion resistance that prevents product contamination
Specifications
Quantity1 Ea.
Length (Metric)150 mm
MaterialSteel
TypeCrucible Tongs
Unit SizeEA